Imprimer

Recombinant protein A, Pierce™

Marques : Thermo Fisher Scientific
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : sur 5

21184 77673 77674
PIER21184EA 202 EUR
PIER21184 PIER77673 PIER77674
Recombinant protein A, Pierce™
Protéines purification Réactifs de purification des protéines

Pierce™ Recombinant Protein A is useful as the basis for preparing various kinds of probes or affinity media for detection or purification of rabbit and human antibodies, especially IgG isotypes, in immunoassays and antibody purification protocols.


  • Contains four Fc-binding domains per protein
  • More efficient than Protein G for polyclonal IgG from rabbit, pig, dog and cat serum
  • Less efficient than Protein G for mouse IgG1, human IgG3 and rat, goat and cow antibodies


Protein A is a cell wall component produced by several strains of Staphylococcus aureus that consists of a single polypeptide chain and contains little or no carbohydrate. Recombinant Protein A is produced in E. coli and functions essentially the same as native Protein A. The Protein A molecule contains four high-affinity binding sites capable of interacting with the Fc region from IgG of several species including human and rabbit. Optimal binding occurs at pH 8,2, although binding is also effective at neutral or physiological conditions (pH 7,0 to 7,6).

Commander maintenant


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR