Imprimer
Carrier proteins, Imject™ Blue Carrier™
Marques : Thermo Fisher Scientific
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
77130 77660
PIER77130EA 269 EUR
PIER77130 PIEA77660
Carrier proteins, Imject™ Blue Carrier™
Protéines extraction Protein Extraction Kits
Imject™ Blue Carrier™ protein is a highly soluble, mollusc-derived hemocyanin that enable simple preparation of effective immunogens with amine- or carboxyl-peptide antigens.
  • High-yield conjugation with each molecule of CCH contains hundreds of primary amines for coupling haptens via EDC or NHS ester crosslinkers
  • Validated quality for the immunogenic carrier protein to ensure uniform chemical structure and solubility
  • Superior solubility and highly immunogenic