Imprimer

Pierce™ Antibody Purification Kits, Chicken IgY Purification Kit

Marques : Thermo Fisher Scientific
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : sur 5

44918
PIER44918EA 364 EUR
PIER44918
Pierce™ Antibody Purification Kits, Chicken IgY Purification Kit
Protéines purification Protein Purification Kits

Pierce™ chicken IgY purification kit purifies chicken antibodies from hen egg yolks using a unique delipidation reagent to separate protein from lipid and a precipitation reagent to selectively isolate the IgY fraction.


  • Purifies twice the amount of IgY and obtain 80 to 120 mg IgY per egg yolk at 85% to 95% purity
  • Simple precipitation method purifies chicken IgY without affinity columns
  • Eggs can be stored in buffer and purified at a later date


Informations de livraison: Kit includes delipidation reagent (500 ml or 5×500 ml), IgY precipitation reagent (500 ml or 5×500 ml), egg separator, (1 or 5 separators).

Commander maintenant


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR