Imprimer
Blocking buffers, protein-free, Pierce™
Marques : Thermo Fisher Scientific
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
37573 37572 37585 37584
PIER37573EA 268 EUR
PIER37573 PIER37572 PIER37585 PIER37584
Blocking buffers, protein-free, Pierce™
Tampons Tampons d'électrophorèse Tampons bloquants
Pierce Protein-free blocking buffer is a non-protein compound that provides effective blocking for membrane-based and plate-based protein detection methods, resulting in extremely low background. It is supplied in a convenient, ready-to-use format.
  • Protein-free blocker - minimises or eliminates cross-reactivity associated with protein-based blocking buffers
  • Applications - compatible with western blotting (membranes), ELISA (microplates), and arrays (coated glass slides)
  • Streptavidin-friendly - free of biotin; no interference with avidin-biotin detection systems
  • High-performance - optimised and validated in many protein methods to provide high signal-to-noise ratio (i.e., no quenching of specific binding and signal but eliminating nonspecific binding and background)
Traditional blocking buffers contain proteins that can cross-react in certain immunodetection systems, resulting in high background and reduced signal. Pierce Protein-free blocking buffers are devoid of protein while remaining highly effective at blocking plates, membranes, and other surfaces for ELISA, western blotting, glass slide arrays, and other applications.
Attention: For research use only. Not for use in diagnostic procedures.