Imprimer
Sample cups for analysers
Fournisseur: PerkinElmer
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
B0510397 B3001566 B3001567 B0087600 B0119079
PERCB0510397EA 320 EUR
PERCB0510397 PERCB3001566 PERCB3001567 PERCB0087600 PERCB0119079
Sample cups for analysers
Récipients de collecte d'échantillon Coupelles pour échantillons d'analyses
Disposable.
These sample cups are of heavy-duty construction to prevent cracking or tipping. Their conical design allows for use of nearly the entire sample. For concentrated acids including HF for analyses.