Imprimer
Presse à granulés hydraulique, KBr
Fournisseur: PerkinElmer
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
51004599
PERC51004599EA 10960 EUR
PERC51004599
Presse à granulés hydraulique, KBr
Presses
This compact hydraulic press is used with the KBr die to prepare KBr pellets.
Pressure is produced by operating a pump lever.
Informations de commande: Evacuable KBr die, KBr pellet holder and adjustable adapter have to be ordered separately.