Imprimer
Wall hooks
Fournisseur: Palbam
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
GJ0401 GJ0501 GJ1001
PALBGJ0401EA 250 EUR
PALBGJ0401 PALBGJ0501 PALBGJ1001
Wall hooks
Crochets muraux
These wall hooks of varying lengths provide a simple solution for hanging lab coats. Made from stainless steel with electropolished finish.