Imprimer
Cell culture media, Minimum Essential Medium (MEM)
Marques : Biowest
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
L0430-500 L0415-500 L0440-500 L0475-500 L0476-500 P0440-N1L P0440-N10L P0450-N1L P0450-N10L P0451-N1L P0451-N10L P0515-N1L P0515-N10L P0440-N5L P0450-N50L P0451-N50L P0515-N50L L0445-500 L0444-500 P0451-N5L L0470-500 P0440-N50L L0416-500
L0430-500EA 10.9 EUR
L0430-500 L0415-500 L0440-500 L0475-500 L0476-500 P0440-N1L P0440-N10L P0450-N1L P0450-N10L P0451-N1L P0451-N10L P0515-N1L P0515-N10L P0440-N5L P0450-N50L P0451-N50L P0515-N50L L0445-500 L0444-500 P0451-N5L L0470-500 P0440-N50L L0416-500
Cell culture media, Minimum Essential Medium (MEM)
Milieux de culture cellulaire
Minimum Essential Medium (MEM) with Earle's Balanced Salts is a modification of Eagle's earlier medium, Basal Medium Eagle (BME), containing higher concentrations of the essential nutrients. These media have demonstrated the ability to support a variety of normal and transformed cells in culture and contain Earle's Balanced Salts, which make them suitable for use in atmospheres charged with CO₂ gas.