Imprimer

X-Gal (Bromo-5-chloro-4-indolyl-3-ß-D-galactopyranoside) kit_staining

Marques : OZ BIOSCIENCES
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : sur 5

Synonyme(s): BCIG

GX10003
OZBIGX10003EA 199.16 EUR
OZBIGX10003
X-Gal (Bromo-5-chloro-4-indolyl-3-ß-D-galactopyranoside) kit_staining
X-Gal (Bromo-5-chloro-4-indolyl-3-ß-D-galactopyranoside)

X-Gal staining kit allows to visualise ß-gal expression through hydrolysis of the X-Gal (5-bromo-4-chloro-3-indoyl-beta-D-Galactopyranoside) substrate yielding blue precipitates.


  • Convenient for all LacZ transfection assays
  • Suitable for cultured cells and tissues
  • Complete kit provided with substrate and buffers
  • Economical


LacZ is one of the most frequently reporter gene used in transfection experiments. The LacZ encoded protein (beta-galactosidase, beta-Gal), is very stable, resistant to proteolytic degradation and easily tested.


Informations de livraison: Kit contains fixing buffer (125 ml), 10X PBS (75 ml), staining buffer (125 ml) and 25X stock solution of X-Gal (4 ml).


Attention: Store at +4 °C except 25X stock solution of X-Gal –20 °C.

Formule: C₁₄H₁₅BrClNO₆
Poids moléculaire: 408.63 g/mol
Numéro MDL: MFCD00005666
Numéro CAS: 7240-90-6

Commander maintenant


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR