Imprimer
DNA transfection reagent, EcoTransfect™
Marques : OZ BIOSCIENCES
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
ET10500 ET11000 ET13000
OZBIET10500EA 111.8 EUR
OZBIET10500 OZBIET11000 OZBIET13000
DNA transfection reagent, EcoTransfect™
Réactifs de transfection
EcoTransfect™ transfection reagent is an economical lipofection reagent dedicated to the transfection of popular cell lines. It is the perfect solution for quickly analysing the biological activity of nucleic acids, to perform routine transfection assays at low cost and to accomplish high throughput screening.
  • Economical
  • Good efficiency
  • Serum compatible
  • Simple, ready to use and rapid
Store at +4 °C.
Informations de livraison: Shipped at room temperature.