Imprimer
DNA transfection reagent, DreamFect™ Gold
Marques : OZ BIOSCIENCES
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
DG80500 DG81000 DG85000
OZBIDG80500EA 200.22 EUR
OZBIDG80500 OZBIDG81000 OZBIDG85000
DNA transfection reagent, DreamFect™ Gold
Réactifs de transfection
DreamFect™ Gold transfection reagent allows transfecting all types of nucleic acids with a very high efficiency. Based on Tee-technology (Triggered Endosomal Escape), DreamFect™ Gold delivers a large quantity of nucleic acids leading to higher protein expression compared to other transfection reagents. It is fully and does not interfere with cellular mechanisms. Consequently, high cell viability is maintained in every experiment and any potential secondary effect is avoided.
  • High efficiency
  • Higher protein expression
  • Superior protein expression level
  • Avoid secondary effects
  • Serum compatible
  • Simple, rapid and easy to use
Store at –20 °C.
Informations de livraison: Shipped at room temperature.