Imprimer
Culti-Loops™ Streptococcus dysgalactiae subsp. equisimilis ATCC™ 35666™
Marques : THERMO FISHER DIAGNOSTICS
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
R4608217
OXOIR4608217EA 342 EUR
OXOIR4608217
Culti-Loops™ Streptococcus dysgalactiae subsp. equisimilis ATCC™ 35666™
Tests microbiologiques Tests sur les micro-organismes
Culti-Loops™ are ready-to-use QC organisms recommended for use in performance testing of media, stains, reagents and identification kits, and for the evaluation of bacteriological procedures.
The loop may be dissolved in liquid medium or streaked directly onto an appropriate media.
Cdt: Culti-Loops are individually sealed in a foil pouch and packaged five loops per package.