Imprimer
Culti-Loops™ Staphylococcus aureus subsp. aureus ATCC™ 49476™
Marques : THERMO FISHER DIAGNOSTICS
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
R4606510
OXOIR4606510EA 342 EUR
OXOIR4606510
Culti-Loops™ Staphylococcus aureus subsp. aureus ATCC™ 49476™
Tests microbiologiques Tests sur les micro-organismes
Culti-Loops™ are ready-to-use QC organisms recommended for use in performance testing of media, stains, reagents and identification kits, and for the evaluation of bacteriological procedures.
The loop may be dissolved in liquid medium or streaked directly onto an appropriate media.
Cdt: Culti-Loops are individually sealed in a foil pouch and packaged five loops per package.