Imprimer
Culti-Loops™ Haemophilus influenzae Type A ATCC™ 9006™
Marques : THERMO FISHER DIAGNOSTICS
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
R4603805
OXOIR4603805EA 342 EUR
OXOIR4603805
Culti-Loops™ Haemophilus influenzae Type A ATCC™ 9006™
Tests microbiologiques Tests sur les micro-organismes
Culti-Loops™ are ready-to-use QC organisms recommended for use in performance testing of media, stains, reagents and identification kits, and for the evaluation of bacteriological procedures.
The loop may be dissolved in liquid medium, or streaked directly onto an appropriate media.
Cdt: Culti-Loops are individually sealed in a foil pouch and packaged five loops per package.