Imprimer
Culti-Loops™ Klebsiella pneumoniae subsp. pneumoniae ATCC™ 700603™
Marques : THERMO FISHER DIAGNOSTICS
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
R4603074
OXOIR4603074EA 342 EUR
OXOIR4603074
Culti-Loops™ Klebsiella pneumoniae subsp. pneumoniae ATCC™ 700603™
Tests microbiologiques Tests sur les micro-organismes
Culti-Loops™ are ready-to-use QC organisms recommended for use in performance testing of media, stains, reagents and identification kits, and for the evaluation of bacteriological procedures.
The loop may be dissolved in liquid medium or streaked directly onto an appropriate media.
Cdt: Culti-Loops are individually sealed in a foil pouch and packaged five loops per package.