Imprimer
Culti-Loops™ Serratia odorifera ATCC™ 33077™
Marques : THERMO FISHER DIAGNOSTICS
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
R4602000
OXOIR4602000EA 342 EUR
OXOIR4602000
Culti-Loops™ Serratia odorifera ATCC™ 33077™
Tests microbiologiques Tests sur les micro-organismes
Culti-Loops™ are ready-to-use QC organisms recommended for use in performance testing of media, stains, reagents and identification kits, and for the evaluation of bacteriological procedures.
The loop may be dissolved in liquid medium or streaked directly onto an appropriate media.
Cdt: Culti-Loops are individually sealed in a foil pouch and packaged five loops per package.
Commander maintenant
SPÉCIFICATIONS
Environmentally Preferable