Imprimer
Bacterial antigen rapid latex agglutination test, Wellcogen™
Marques : THERMO FISHER DIAGNOSTICS
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
R30858701 R30858801 R30859001 R30859203 R30859502 R30859602 R30164501 R30368601
OXOIR30858701EA 332 EUR
OXOIR30858701 OXOIR30858801 OXOIR30859001 OXOIR30859203 OXOIR30859502 OXOIR30859602 OXOIR30164501 OXOIR30368601
Bacterial antigen rapid latex agglutination test, Wellcogen™
Tests microbiologiques Tests sur les anticorps Ag-AK, Latex
Directly detect antigens present in human cerebrospinal fluid (CSF), serum, urine, or blood cultures using Thermo Scientific™ Wellcogen™ Bacterial Antigen Rapid Latex Agglutination Test.
  • Not all products are available for sale in all territories