Imprimer
pH electrode, double junction, Orion™ 9102DJWP
Fournisseur: Thermo Orion
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
9102DJWP
ORIO9102DJWPEA 197 EUR
ORIO9102DJWP
pH electrode, double junction, Orion™ 9102DJWP
Électrodes Électrodes pH
This double junction pH electrode offers superior measurement stability, faster response, greater accuracy, longer life, least long-term drift and more reproducible results.
  • With waterproof BNC connector
  • Ag/AgCl double junction
  • Refillable