Imprimer
Human Recombinant CDC2 (His Tag) (from E. coli)
Marques : Novus Biologicals
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
NBP2-52017-0.1
NOVUNBP2-52017-0.1EA 486 EUR
NOVUNBP2-52017-0.1
Human Recombinant CDC2 (His Tag) (from E. coli)
Protéines et Peptides
A denatured recombinant protein with a N-terminal his-tag and corresponding to the amino acids 1 - 297 of human CDC2/CDK1.