Imprimer

Anti-SERF1A Rabbit Polyclonal Antibody (DyLight® 405)

Fournisseur: Novus Biologicals
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : sur 5

NBP1-78393V
NOVUNBP1-78393VEA 668 EUR
NOVUNBP1-78393V
Anti-SERF1A Rabbit Polyclonal Antibody (DyLight® 405)
Anticorps
Type: Primary
Antigen: SERF1A (small EDRK-rich factor 1A (telomeric))
Clonality: polyclonal
Clone:
Conjugation: DyLight® 405
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR