Imprimer

Anti-Ethenoadenosine Mouse Monoclonal Antibody (DyLight® 650) [clone: 1G4]

Fournisseur: Novus Biologicals
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : sur 5

NB600-442C
NOVUNB600-442CEA 710 EUR
NOVUNB600-442C
Anti-Ethenoadenosine Mouse Monoclonal Antibody (DyLight® 650) [clone: 1G4]
Anticorps
Type: Primary
Antigen: Ethenoadenosine
Clonality: monoclonal
Clone: 1G4
Conjugation: DyLight® 650
Epitope:
Host: Mouse
Isotype: IgG2 Lambda
Reactivity: Pan
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR