Imprimer

Anti-TH Rabbit Polyclonal Antibody

Fournisseur: Novus Biologicals
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : sur 5

NB300-109
NOVUNB300-109EA 764 EUR
NOVUNB300-109
Anti-TH Rabbit Polyclonal Antibody
Anticorps
Rabbit Polyclonal Tyrosine Hydroxylase Antibody. Tested Applications: Western Blot, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry-Frozen. Tested Reactivity: Human, Mouse, Rat, Mammal.

There are 2 scientific publications using this antibody.

Type: Primary
Antigen: Tyrosine Hydroxylase
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR