Imprimer

Anti-MNAT1/MAT2A Rabbit Polyclonal Antibody (DyLight® 488)

Fournisseur: Novus Biologicals
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : sur 5

NB110-94162G
NOVUNB110-94162GEA 722 EUR
NOVUNB110-94162G
Anti-MNAT1/MAT2A Rabbit Polyclonal Antibody (DyLight® 488)
Anticorps
Rabbit Polyclonal MAT1/2A Antibody [DyLight 488]. Tested Applications: Western Blot. Tested Reactivity: Human, Rat, Bovine, Primate, Zebrafish, Mouse (-).

Type: Primary
Antigen: MNAT1/MAT2A
Clonality: Polyclonal
Clone:
Conjugation: DyLight® 488
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR