Imprimer

Anti-CD4 Rabbit Polyclonal Antibody

Fournisseur: Novus Biologicals
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : sur 5

NB100-56457
NOVUNB100-56457EA 596 EUR
NOVUNB100-56457
Anti-CD4 Rabbit Polyclonal Antibody
Anticorps
Type: Primary
Antigen: CD4 (CD4 molecule)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR