Imprimer

Anti-CDK5R1 Rabbit Polyclonal Antibody

Fournisseur: Novus Biologicals
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : sur 5

NB100-56153
NOVUNB100-56153EA 516 EUR
NOVUNB100-56153
Anti-CDK5R1 Rabbit Polyclonal Antibody
Anticorps
Type: Primary
Antigen: CDK5R1 (cyclin-dependent kinase 5, regulatory subunit 1 (p35))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Pan
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR