Imprimer
Bowls, evaporating, with spout
Fournisseur: NICKEL ELECTRO
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
715
NICK715EA 69.8 EUR
NICK715
Bowls, evaporating, with spout
Plateaux/Récipients/Pots Capsules d'évaporation
Pure nickel (99,6%).
  • Excellent corrosion and temperature resistance
  • Smooth surfaces for fast, easy cleaning
Round, with flat bottom. Multipurpose tray ideal for a wide variety of uses.