Imprimer
Cinchable leak diverter, PIG®
Fournisseur: NEW PIG
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
TLS182
NEWPTLS182EA 71.3 EUR
NEWPTLS182
Cinchable leak diverter, PIG®
Matériaux absorbants
The leak diverter made from PVC/polyester features drawstrings which cinch tightly around valves, fittings and other oddly shaped components to capture leaks at the source.
  • Compact size for leaks in tight spaces
  • Non-permeable material is vinyl-coated for strength and durability
  • Resists punctures and mildew for long life
  • UV resistant up to 12 months for long-term outdoor use without degradation
Accepts a standard 3/4" hose connection to safely channel liquid to floor drain or collection container (hose sold separately). Flow rate of 12,6 litres perEn savoir plus