Imprimer
Whirl-Pak® Cellulose Sponge Bags
Fournisseur: NASCO FORT ATKINSON
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
B01245 B01423 B01422
NASCB01245EA 480 EUR
NASCB01245 NASCB01423 NASCB01422
Whirl-Pak® Cellulose Sponge Bags
Sachets Sachets d'échantillonnage
Cellulose sponges for your environmental monitoring and surface sampling needs.
  • Plus 1 sterilisation, post-manufacturing sterilisation: Increased sample integrity, perforated closure to maintain sterility
  • Convenient write-on strip for easy labelling
  • Puncture-proof closure tabs: Inner and outer tapes for longevity and stability, wire for easy opening, pouring, and closing
  • Opening pull tabs: For easy opening, non-slip textured Tyvek
  • Continuous perimeter seal: Maximum protection against leaks
  • Tape extends beyond wire to eliminate sharp ends and provide a
En savoir plus