Imprimer
Papiers pour laboratoire, Nalgene®
Fournisseur: Thermo Fisher Scientific
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
6304-9811
NALG6304-9811EA 112 EUR
NALG6304-9811
Papiers pour laboratoire, Nalgene®
Papiers
These polyolefin sheets roll or fold without cracking and stay flexible at cold temperatures. They won't rip, stretch or shrink. They can be punched for use in binders or cut into labels. These sheets do not accept pencil.
  • Resistant to many solvents including acetone, formaldehyde, formalin, xylene, acetic acid and ethanol
  • Accept many inks including laser, inkjet, copier, markers, ballpoint, gels and technical markers
Format : A4