Imprimer

Bleu nitré-tétrazolium chlorure (NBT) 90 HPLC_ASSAY_METHOD, Sigma-Aldrich®

Marques : SIGMA ALDRICH MICROSCOPY
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : sur 5

Attention

Synonyme(s): 5,5'-Diphenyl-3,3'-bis(4-nitrophenyl)-2,2'-(3,3'-dimethoxybiphenyl-4,4'-ylene)ditetrazolium dichloride

MQ200

N6876-1G N6876-100MG N6876-10X10MG N6876-250MG N6876-500MG N6876-50MG N6876-5G
N6876-1GEA 328 EUR
N6876-1G N6876-100MG N6876-10X10MG N6876-250MG N6876-500MG N6876-50MG N6876-5G
Bleu nitré-tétrazolium chlorure (NBT) 90 HPLC_ASSAY_METHOD, Sigma-Aldrich®
Bleu nitré-tétrazolium chlorure (NBT)

Nitrotetrazolium blue (NBT), in combination with BCIP (5-bromo-4-chloro-3-indolyl-phosphate), is required for the detection of labeled probes in western blots. It is part of the colorimetric detection assay. NBT and BCIP dyes detect alkaline phosphatase activity, thereby generating an insoluble purple dye (diformazan). The dye is visible at areas where the labeled probe has hybridised to its target.


  • Nitrotetrazolium Blue chloride has been used to detect the generation of oxygen metabolites in cells
  • It has been used as a substrate for western blot detection
  • It has been used for the staining of histological tissue sections
  • Substrate for dehydrogenases and other oxidases

Formule: C₄₀H₃₀Cl₂N₁₀O₆
Poids moléculaire: 817.65 g/mol
Point de fusion: 200
Température de stockage: Réfrigérateur
Numéro MDL: MFCD00012159
Numéro CAS: 298-83-9
EINECS: 206-067-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9de2c2cec7b7e51c50562030878bfa557af5df16&vid=175a4c932ab1f09b9b2613c727f5a3dadf3be4b0&prid=web&clid=SAPDAM

Commander maintenant

Résultats des tests de spécification

Appearance (Colour) Yellow
Appearance (Form) Powder
Infrared spectrum Conforms to Structure
Water (by Karl Fischer) ≤4.5 %
Solvent Content (by GC) ≤10.0 %
Nitrogen
Corrected for Water and Solvent
16.4 - 17.9 %
Purity (HPLC) ≥90.0 %
Solubility (Colour)
c = 10 mg/ml; Water
Green-Yellow to Dark Yellow
Solubility (Turbidity) Clear
Recommended Retest Period 3 Years


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR