Imprimer

Supelco® Cerilliant® Organic Reference Standard, (±)-Norepinephrine-D₆ hydrochloride solution

Marques : Merck
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : sur 5

N-069-1ML
N-069-1MLEA 173 EUR
N-069-1ML
Supelco® Cerilliant® Organic Reference Standard, (±)-Norepinephrine-D₆ hydrochloride solution
Étalons Étalons organiques

  • GHS signal: GHS02, GHS06, GHS08


Attention: P210 - P280 - P301 + P310 + P330 - P302 + P352 + P312 - P304 + P340 + P311

https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=143243c0c1997a3fd4e1b8843ddefe777b818c71&vid=143243c0c1997a3fd4e1b8843ddefe777b818c71&prid=web&clid=SAPDAM

Commander maintenant

SPÉCIFICATIONS

Environmentally Preferable
M-Clarity quality level MQ300
N ° CAS. 1219803-04-9
Molecular weight 211,68
Description de la classe de danger H225 - H301 + H311 + H331 - H370
Point d'éclair 9,7 °C - Closed cup
Conditions de transport DICE
Conditions de stockage –70 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR