Imprimer
Safety Storage Cabinet, Type 90, ION-CORE-90 CHARGE model IO90.195.060.CC.WDC
Fournisseur: asecos
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
41371 41373 41376 41302047 41366 41369
MSGA41371EA 0 EUR
MSGA41371 MSGA41373 MSGA41376 135-2885 MSGA41366 MSGA41369
Safety Storage Cabinet, Type 90, ION-CORE-90 CHARGE model IO90.195.060.CC.WDC
Mobilier Armoires de stockage
Safe active and passive storage of lithium-ion batteries.
  • Robust construction and longevity
  • Integrated potential-free alarm contact
  • No unauthorised use
  • Integrated transport base
  • Easy alignment
  • Smoke detector
  • Pressure relief
  • All-around protection
Robust construction and longevity: robust, high-quality, scratch-resistant outer body with powder-coated textured surface. Integrated potential-free alarm contact for strongly recommended alarm forwarding. Optionally available with module for remote signalling.
No unauthorised use: door lockable withEn savoir plus
Commander maintenant
SPÉCIFICATIONS
Environmentally Preferable