Imprimer
Safety cabinets for chemicals, CS-Classic
Fournisseur: asecos
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
30557050 30561040 30561041 30561046 30561047 30561048 30561049 30561050 30562040 30562041 30562046 30562047 30562048 30562049 30562050 30563040 30563041 30563046 30563047 30563050 30564040 30564041 30564046 30564047 30564048 30564049 30564050 3055704130565
MSGA30557050EA 651 EUR
MSGA30557050 MSGA30561040 MSGA30561041 MSGA30561046 MSGA30561047 MSGA30561048 MSGA30561049 MSGA30561050 MSGA30562040 MSGA30562041 MSGA30562046 MSGA30562047 MSGA30562048 MSGA30562049 MSGA30562050 MSGA30563040 MSGA30563041 MSGA30563046 MSGA30563047 MSGA30563050 MSGA30564040 MSGA30564041 MSGA30564046 MSGA30564047 MSGA30564048 MSGA30564049 MSGA30564050 MSGA3055704130565
Safety cabinets for chemicals, CS-Classic
Mobilier Armoires de stockage
Safety storage cabinets for non-flammable chemicals and toxic substances in workrooms.
  • Body made of sheet steel with high grade structured paint finish with robust construction and longevity
  • Lockable doors with cylinder locking integrated in the turning handle
  • Adusting aids to compensate for uneven floor
  • Natural ventilation due to vent openings at the cabinet bottom for the reduction of odours
Informations de commande: Supplied without interior equipment packages. Interior equipment packages need to be ordered in addition. Please check the website orEn savoir plus