Imprimer
Safety storage cabinets, Type 30, K-PHOENIX-30
Fournisseur: asecos
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
30343001
MSGA30343001EA 3770 EUR
MSGA30343001
Safety storage cabinets, Type 30, K-PHOENIX-30
Mobilier Armoires de stockage
Safe and approved storage of aggressive and flammable hazardous substances in working areas.
  • Robust construction and longevity - triple hinged door, body made of powder-coated sheet steel, safety elements assembled outside the cabinet for increased protection against corrosion
  • Two in one - 2 compartments (vertical separation) allow the storage of flammable liquids and aggressive hazardous materials in one cabinet
  • User-friendly and safe (folding door compartment): door open arrest system (standard), convenient and safe removal or storage of containers
  • No
En savoir plus