Imprimer

Dextran sulphate sodium salt

Marques : Molekula
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : sur 5

Synonyme(s): Dextran sulphate sodium from Dextran

38417956-25G
MOLE38417956-25GEA 0 EUR
MOLE38417956-25G
Dextran sulphate sodium salt
Dextran sulphate sodium salt
Formule: (C₆H₇Na₃O₁₄S₃)n
Numéro MDL: MFCD00081551
Numéro CAS: 9011-18-1

Commander maintenant


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR