Imprimer
Pura-Fit® Classic Foam Earplugs
Fournisseur: Moldex
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
776001
MOLD776001EA 130 EUR
MOLD776001
Pura-Fit® Classic Foam Earplugs
Bouchons d'oreille Bouchons d'oreilles à usage unique
The classic Pura-Fit® earplugs offer the optimal combination of stability and softness. They are easy to roll and, upon insertion, expand to fill the ear canal at exactly the right moment. There, they maintain a secure, sealed fit, without exerting pressure on the ear.
  • Noise reduction at high (H), medium (M) and low (L) frequencies
  • 36 dB (SNR) noise reduction
  • Soft foam for a high degree of wearing comfort
  • Also available in a dispenser and with neck cord
Shelf life from date of production: 4 years.