Imprimer
Molecular detection assay E. coli O157
Fournisseur: 3M Food Safety
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
MDA2ECO96
MMMPMDA2ECO96EA 1160 EUR
MMMPMDA2ECO96
Molecular detection assay E. coli O157
Systèmes de surveillance Tests de contrôle de l'hygiène
3M™ Molecular Detection Assay 2 - E. coli O157 (including H7) is used with the 3M™ Molecular Detection System for the rapid and specific pathogen detection of E. coli O157, including H7, in enriched food samples.
  • One assay protocol and fewer steps than most tests reduces repetitive tasks and makes it easy to train employees
  • Ready-to-use and pre-dispensed reagents provide convenience and minimise the chance of human error
  • Color-coded assay tubes by organism and matching software screens allow for better tracking and easy identification
  • The ability to run
En savoir plus