Imprimer
CloneMatrix
Marques : Molecular Devices
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
K8510 K8530
MLDVK8510EA 249 EUR
MLDVK8510 MLDVK8530
CloneMatrix
Milieux de culture cellulaire
CloneMatrix is a 2,5X concentrate that serves as a matrix to produce semi-solid media for hybridoma, myeloma, CHO, HEK, and mESC.