Imprimer
Microplate readers, multi-mode, SpectraMax® Mini
Fournisseur: Molecular Devices
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
SMAXMINIAFL SMAXMINIAF SMAXMINIAL 5089096 5089097 5089098 5089099 5089100 5089102 5089103 5089104 5089334
735-0396EA 0 EUR
735-0396 735-0395 735-0397 MLDV5089096 MLDV5089097 MLDV5089098 MLDV5089099 MLDV5089100 MLDV5089102 MLDV5089103 MLDV5089104 MLDV5089334
Microplate readers, multi-mode, SpectraMax® Mini
Lecteurs de microplaques
The SpectraMax® Mini multi-mode microplate reader offers a budget friendly solution that gives you exactly what you need when you need it. With three modes of detection for UV/Vis absorbance, fluorescence and luminescence, the SpectraMax® Mini reader is user-upgradeable - from two to three modes, providing researchers with a versatile instrument that does not compromise on sensitivity or performance.
  • Industry leading software - enjoy an extensive library of protocols, advanced reader control, and easy data management and analysis with our 'best in class'
En savoir plus