Imprimer
Sterivex™ Membrane Filter Units, with Durapore® (PVDF) Membrane
Fournisseur: Merck
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
SVGV01015 SVGVB1010 SVHVB1010 SVHV01015 SVHVL10RC SVGV010RS SVGVL10RC SVHV010RS
MILFSVGV01015EA 248 EUR
MILFSVGV01015 MILFSVGVB1010 MILFSVHVB1010 MILFSVHV01015 MILFSVHVL10RC MILFSVGV010RS MILFSVGVL10RC MILFSVHV010RS
Sterivex™ Membrane Filter Units, with Durapore® (PVDF) Membrane
Filtres Unités à membrane filtrante
The Sterivex™ filter unit has a compact design ideal for large volume filtration using positive pressure.
  • Filters can fit virtually any workflow
  • Filters dispense into nearly any storage container
The filter units work with syringes, peristaltic pumps or pressure vessels, thus eliminates the risk of contamination associated with decanting or pipetting. Filter-sterilise volumes up to 1000 ml or 2000 ml, using pressure driven filter units (capacity depends on the type of membrane used in the device).