Imprimer
Aerosol Analysis Monitors, 37 mm
Fournisseur: Merck
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
MAWP037A0 MAWP037AM MHWP037A0
MILFMAWP037A0EA 326 EUR
MILFMAWP037A0 MILFMAWP037AM MILFMHWP037A0
Aerosol Analysis Monitors, 37 mm
Filtres Filtres pour cassettes
Millipore's 37 mm Aerosol analysis monitors are transparent, disposable, plastic filter holders pre-assembled with Type MF (mixed cellulose esters) Millipore filters. A thin cellulose support seals the filter between monitor sections to distribute sample flow evenly over the filter surface. The three-part design holds the filter in-place when the top section is removed for open aerosol sampling.
  • Pre-assembled with filters in-place
  • Available with matched-weight membranes
The Average Background particle Count (ABC) value is marked on each monitorEn savoir plus