Imprimer
Membrane filters, Durapore®
Fournisseur: Merck
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
GVWP04700 GVWP09050 HVLP04700 SVLP04700 VVHP04700 GVWP01300 DVPP01300 DVPP02500 DVPP04700 DVPP09050 DVPP14250 GVHP01300 GVHP02500 GVHP09050 GVHP14250 GVWP10050 HVHP02500 HVHP04700 HVHP09050 HVHP14250 HVLP01300 HVLP14250 VVLP04700 HVLP09050 SVLP01300 SVLP02500 SVLP09050 SVWG04700 VVLP01300 VVLP02500 VVLP09050 VVLP14250 GVWP14250 GVHP04700 GVWP02500 HVLP02500 GVHP00010 HVHP01300 VVLP00010 DVPP00010 GVWP00010 DVPP08250 HVLP00010 HVWG04700 SLVP07550 DVPP29325 HVLP08050
512-0254EA 375 EUR
512-0254 MILFGVWP09050 512-0262 MILFSVLP04700 MILFVVHP04700 588-4110 MILFDVPP01300 MILFDVPP02500 MILFDVPP04700 MILFDVPP09050 MILFDVPP14250 MILFGVHP01300 MILFGVHP02500 MILFGVHP09050 MILFGVHP14250 MILFGVWP10050 MILFHVHP02500 512-0261 MILFHVHP09050 MILFHVHP14250 MILFHVLP01300 MILFHVLP14250 MILFVVLP04700 512-0263 MILFSVLP01300 MILFSVLP02500 MILFSVLP09050 MILFSVWG04700 MILFVVLP01300 MILFVVLP02500 MILFVVLP09050 MILFVVLP14250 MILFGVWP14250 MILFGVHP04700 MILFGVWP02500 MILFHVLP02500 MILFGVHP00010 MILFHVHP01300 MILFVVLP00010 MILFDVPP00010 MILFGVWP00010 MILFDVPP08250 MILFHVLP00010 MILFHVWG04700 MILFSLVP07550 MILFDVPP29325 MILFHVLP08050
Membrane filters, Durapore®
Filtres Filtres à membrane
Durapore® PVDF white membranes provide high flow rates and throughput, low extractables and broad chemical compatibility. Hydrophlic membrane binds far less protein than nylon, nitrocellulose, or PTFE membranes.
  • Can be sterilised by ethylene oxide and gamma irradiation
  • Available in several pore sizes (both hydrophilic and hydrophobic)
  • Have very low protein binding to minimize interaction with your sample and maximise recovery
Filter surface: plain
Bacterial endotoxins: 0,5 EU/ml
Gravimetric extractables: <0,5%