Imprimer
Durapore® Charged Membrane Filter
Fournisseur: Merck
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
CCGL09025
MILFCCGL09025EA 524 EUR
MILFCCGL09025
Durapore® Charged Membrane Filter
Filtres Filtres à membrane
Durapore® membranes provide high flow rates and throughput, low extractables and broad chemical compatibility.
Charged hydrophlic Durapore® membrane (CCGL) has a positive net charge which allows the membrane disc to separate negatively charged particles that are smaller than the rated pore size of the disc from the liquid solution.