Imprimer

Hématoxyline solution_SC (Mayer), Sigma-Aldrich®

Marques : SIGMA ALDRICH MICROSCOPY
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : sur 5

Attention

Synonyme(s): DcH-48, Natural Black I, Hydroxybrasilin

MHS1-100ML
MHS1-100MLEA 47.2 EUR
MHS1-100ML
Hématoxyline solution_SC (Mayer), Sigma-Aldrich®
Hématoxyline (solution de coloration)

General purpose nuclear stain, progressive type. Used with hematoxylin and eosin staining.

Formule: C₁₆H₁₄O₆
Poids moléculaire: 302.28 g/mol
Température de stockage: Température ambiante
Numéro MDL: MFCD00078111
Numéro CAS: 517-28-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=577c852c1cc9e5db58ddeb207f71709947345357&vid=e8dacf6ca65863a2c7e2e6f1698778e7bded4caf&prid=web&clid=SAPDAM IVDLogo

Commander maintenant


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR