Imprimer
Burkert® Micro Solenoid Rocker Valves
Fournisseur: Avantor Fluid Handling
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
98622-00
MFLX98622-00EA 490 EUR
MFLX98622-00
Burkert® Micro Solenoid Rocker Valves
Vannes Solenoid Valves
No worrying about reliability—longest life available.
  • Eliminates heat transfer from diaphragm to actuator
  • Easy purging—minimal internal volume
  • Ideal for manifold mounting
Rocker Valves have a miniature rocker that operates the isolated diaphragm to separate actuator from the media. Valve is only 16 mm wide. Each valve is equipped with an LED display and push-on tube connections. Valves include two 19" electrical leads.