Imprimer
Masterflex® I/P® Bulk-Packed Precision Pump Tubing, Platinum-Cured Silicone, Avantor®
Fournisseur: Avantor Fluid Handling
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
MFLX96404-26EA 2470 EUR
MFLX96404-26
Masterflex® I/P® Bulk-Packed Precision Pump Tubing, Platinum-Cured Silicone, Avantor®
Tuyaux Tubes pour pompe péristaltique
Reduce the risk of particulate entrapment with smooth inner surface.
  • Genuine Masterflex® tubing undergoes robust testing to provide optimal performance/pump life hours with peristaltic pump equipment
  • Precision extruded to tight tolerances for high repeatability, accuracy, and reproducibility
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimize downtime
  • Regulatory Approvals to match your unique application
  • Always in stock—most formulations and sizes
En savoir plus