Imprimer
Masterflex® L/S® Digital Modular Dispensing Pump Systems, Avantor®
Fournisseur: Avantor Fluid Handling
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
MFLX77923-80-EUEA 5370 EUR
MFLX77923-80-EU MFLX77923-90-EU
Masterflex® L/S® Digital Modular Dispensing Pump Systems, Avantor®
Pompe Pompes péristaltiques
Precision digital drives offer three dispensing modes and precision low-maintenance motors— modular format lets you place motor and controller where convenient.
  • Digital interface with graphical LCD shows pump performance and operating mode— continuous run, timed dispense, copy dispense, or volume dispense
  • Remote control available using analog communication—voltage, current and contact closure
  • Select your preferred language from the user-friendly menu for programming and operation
  • Dispense by Volume in ml (0.001 to 9,999 mL), L, or gal; Copy—1 to 9,999
En savoir plus