Imprimer
Masterflex® Miniflex® OEM Panel Mount Pumps Systems
Fournisseur: MASTERFLEX PROPRIETARY
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
MFLX77222-00EA 1459 EUR
MFLX77222-00 MFLX77222-02 MFLX77222-10 MFLX77222-12 MFLX77222-14
Masterflex® Miniflex® OEM Panel Mount Pumps Systems
Pompe Pompes péristaltiques
Designed for easy integration into OEM designs wherever precision fluid handling is needed.
  • Precise flowrates up from 0,12 to 390 ml/min (3 to 300 min⁻¹)
  • Rugged sturdy design with durable pump head materials
  • Pump head accepts continuous lengths of tubing for clean flow paths
  • Automatic tubing retention makes tube loading quick and easy
  • IP65 Ingress protection; removable head for spray and wipe down cleanability
  • High efficiency and dependable brushless DC motor
  • I/O control features include analogue speed control, direction, tach output, open head sensor
En savoir plus