Imprimer
Masterflex® L/S® Easy-Load® II Pump Heads for High-Performance Precision Tubing, Avantor®
Fournisseur: Avantor Fluid Handling
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
MFLX77200-32EA 824 EUR
MFLX77200-32 MFLX77200-52 MFLX77200-62 MFLX77201-62
Masterflex® L/S® Easy-Load® II Pump Heads for High-Performance Precision Tubing, Avantor®
Pompe Pump Heads Peristaltic Pump Heads
Automatic retention holds tubing securely in place with no manual adjustment—just load the tubing and start the pump.
  • New design and automatic tubing retention allow for multiple tubing sizes—change tubing without removing head from drive
  • Four-roller rotor improves pressure performance and mechanical stability while reducing pulsation
  • Model available with integrated open-head sensor–connects directly to sensor-equipped drives
  • Pump heads are stackable for multichannel applications
  • Compact design for efficient use of space
  • Suitable for use with L/S® High-
En savoir plus