Imprimer
ITT Jabsco High-Purity Air-Operated Double-Diaphragm Pumps
Fournisseur: Avantor Fluid Handling
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
74020-00
MFLX74020-00EA 3320 EUR
MFLX74020-00
ITT Jabsco High-Purity Air-Operated Double-Diaphragm Pumps
Pompe Pompes pneumatiques
Unique air valve ensures a positive shift every time – pump will not stall.
  • All diaphragms, liquid chambers, and manifolds are constructed
  • of 100% virgin PTFE for long life and high purity
  • Unique valve requires no lubrication or prepacking.
  • Easily access air valve without disassembling the pump
Pumps are manufactured under class 1000 clean room conditions to guarantee purity.